missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
Specifications
Density | 2.607 g/mL |
Assay Percent Range | 98% |
Quantity | 100 g |
UN Number | UN1551 |
Solubility Information | Soluble in water. Insoluble in ethanol. |
Formula Weight | 613.82 (anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Antimony potassium tartrate hydrate |
Safety and Handling
Product Identifier
- Potassium antimonyl tartrate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'