Learn More
Alfa Aesar™ Antimony(III) sulfide, 99.5% (metals basis)
Brand: Alfa Aesar™ 041782.09
Code : NEW
Additional Details : CAS Number : 1345-04-6 Weight : 0.01000kg Transport : UN number : 1549 Chem class : 1 Pack group : III
Quantity | 10g |
---|
Chemical Identifiers
Antimony(III) sulfide | |
339.68 | |
UN1549 |
1345-04-6 | |
MFCD00011217 | |
14,713 |
Specifications
Antimony(III) sulfide | |
Pieces | |
(metals basis) | |
MFCD00011217 | |
14,713 | |
550°C |
1345-04-6 | |
99.5% | |
339.68 | |
UN1549 | |
25mm & down | |
10g |