missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) fluoride, 99+%, Thermo Scientific Chemicals
23.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of Freon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 376°C |
Melting Point | 292°C |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN2923 |
Merck Index | 14,709 |
Solubility Information | Soluble in water,methanol and acetone. Insoluble in ammonia. |
Formula Weight | 178.75 |
Percent Purity | ≥99% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'