missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11303948
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
10025-91-9
Cl3Sb
228.11
MFCD00011212
FAPDDOBMIUGHIN-UHFFFAOYSA-K
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
24814
CHEBI:74856
[Cl-].[Cl-].[Cl-].[Sb+3]
This item is not returnable. View return policy
10025-91-9
Cl3Sb
228.11
MFCD00011212
FAPDDOBMIUGHIN-UHFFFAOYSA-K
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
24814
CHEBI:74856
[Cl-].[Cl-].[Cl-].[Sb+3]

Antimony(III) chloride is used as a chlorinating agent, as a fireproofing agent in textiles; in bronzing steel and as a mordant in dyeing as well as a caustic in medicine. It is also used as an apoptosis agent that is used to detect cholesterols and carotenoids. It is also employed to detect certain compounds, such as cholesterols and carotenoids. This agent seems to cause dehydration of cholesterol to 3,5-cholestadiene and 2,4-cholestadiene which produce the blue color seen during the reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 223.5°C
Melting Point 73.4°C
Physical Form Crystalline Aggregates
Quantity 2 g
UN Number UN1733
Merck Index 14,707
Solubility Information Soluble in alcohol and acetone and acids.
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity 99.999%
Grade Puratronic™
Odor Pungent
Sensitivity Moisture sensitive
Density 3.14 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(III) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'