missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99%, Thermo Scientific Chemicals

Catalog Number p-4407244
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7789-61-9
Br3Sb
361.472
MFCD00016317
RPJGYLSSECYURW-UHFFFAOYSA-K
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
24615
tribromostibane
Br[Sb](Br)Br
This item is not returnable. View return policy
7789-61-9
Br3Sb
361.472
MFCD00016317
RPJGYLSSECYURW-UHFFFAOYSA-K
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
24615
tribromostibane
Br[Sb](Br)Br

Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 280°C
Melting Point 96.6°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%
Solubility Information Soluble in alcohol,acetone,ammonia,benzene,chloroform,dilute hydrochloric acid,hydrobromic acid and carbon disulfide.
Formula Weight 361.48
Odor Characteristic
Refractive Index 1.74
Sensitivity Moisture sensitive
Appearance White powder
Density 4.148 g/cm3
Chemical Name or Material Antimony(III) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'