missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF764 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541195
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KKERQREGTG ALEKPDPVAA GSPGLKSPQA PSAGPPYGWE QLSKAPHRGR Sequence homology: Human: 100%
ZNF764 Polyclonal antibody specifically detects ZNF764 in Human samples. It is validated for Western Blot.Specifications
ZNF764 | |
Polyclonal | |
Unconjugated | |
ZNF764 | |
Zinc finger protein 764; ZNF764 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
92595 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96H86 | |
ZNF764 | |
Synthetic peptide directed towards the N-terminal of human ZNF764 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |