missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534350
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is SW480 cell lysate. PA5-34350 can be used with blocking peptide PEP-1393.
YPEL4 Polyclonal antibody specifically detects YPEL4 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
YPEL4 | |
Polyclonal | |
Unconjugated | |
YPEL4 | |
Protein yippee-like 4; yippee like 4; yippee-like 4; yippee-like 4 (Drosophila); YPEL4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219539, 241525 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q65Z93, Q96NS1 | |
YPEL4 | |
A 12 amino acid synthetic peptide near the amino terminus of human YPEL4 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |