missing translation for 'onlineSavingsMsg'
Learn More

XCL1/Lymphotactin Mouse anti-Human, Clone: 109001, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB6951

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441606

  • £337.93 / 500µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

XCL1/Lymphotactin Monoclonal antibody MAB6951 Specifically detects XCL1/Lymphotactin in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

XCL1/Lymphotactin
109001
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P47992
ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human XCL1/Lymphotactin in ELISAs and Western blots. In sandwich ELISAs, less than 0.15% cross-reactivity was observed with recombinant mouse MCP-5, recombinant human (rh) MIP-3 beta , rhGRO alpha , rhTGF-alpha , rhGRO gamma , rhActivin RII Fc Chimera, rhIGF-II, or rhIL-15.
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
XCL1
E. coli-derived recombinant human XCL1/Lymphotactin Val22-Gly114 Accession # P47992
Protein A or G purified from ascites
RUO
Primary
6375
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only