Learn More
wnvNS3 Protease NS2b Cofactor Mouse anti-Viral, Clone: 345712, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB29071
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439775
  • £249.49 / 100µg
Estimated Shipment
27-06-2019
Description
Description
wnvNS3 Protease NS2b Cofactor Monoclonal antibody MAB29071 Specifically detects wnvNS3 Protease NS2b Cofactor in Viral samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications
Specifications
wnvNS3 Protease NS2b cofactor
345712
Unconjugated
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
E. coli-derived recombinant wnvNS3 Protease protein Ser1423-Lys1470, Gly1506-Leu1689 Accession # ABD85079
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
912267
Reconstitute at 0.5 mg/mL in sterile Tris-buffered saline, pH 7.3.
Western Blot, Immunoprecipitation, Neutralization
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 15 ug/mL
ABD85079
Mouse
IgG3
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects West Nile Virus NS2b cofactor of the NS3 Protease complex in direct ELISAs and Western blots.
Virus