missing translation for 'onlineSavingsMsg'
Learn More

Wnt-8a Rat anti-Mouse, Clone: 254126, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2248

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449295

  • £376.32 / 500µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Wnt-8a Monoclonal antibody MAB2248 Specifically detects Wnt-8a in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Wnt-8a
254126
Western Blot 1 ug/mL
Q64527
protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Wnt-8a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Wnt-1, -3a, -4, -5a, or -10b is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WNT8A
E. coli-derived recombinant mouse Wnt-8a Ala20-Trp354 (predicted) Accession # Q64527
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7478
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only