Learn More
Wnt-4 Rat anti-Mouse, Clone: 55010, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB475-500
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450706
  • £427.77 / 500µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Wnt-4 Monoclonal antibody MAB475500 Specifically detects Wnt-4 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Wnt-4
55010
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WNT4
E. coli-derived recombinant mouse Wnt-4 aa 37-76 and 222-295 Accession # P22724
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
54361
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
P22724
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, Wnt4, WNT-4
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Wnt-4 in direct ELISAs and Western blots.