Learn More
WISP-1/CCN4 Mouse anti-Human, Clone: 213609, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB16271
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438785
  • £337.93 / 500µg
Estimated Shipment
09-07-2019
Description
Description
WISP-1/CCN4 Monoclonal antibody MAB16271 Specifically detects WISP-1/CCN4 in Human samples. It is validated for ELISA Capture
Specifications
Specifications
WISP-1/CCN4
213609
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WISP1
Mouse myeloma cell line NS0-derived recombinant human WISP-1/CCN4 Thr23-Asn367 Accession # O95388
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8840
Sterile PBS to a final concentration of 0.5 mg/mL.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O95388
CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human WISP-1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Biglycan, rhDecorin, rhNOV, or rhWISP-3 was observed.