missing translation for 'onlineSavingsMsg'
Learn More

WIF-1 Rat anti-Mouse, Clone: 134714, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB135-500

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438575

  • £337.93 / 500µg

Estimated Shipment date
from Supplier 24-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

WIF-1 Monoclonal antibody MAB135500 Specifically detects WIF-1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

WIF-1
134714
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WIF1
Mouse myeloma cell line NS0-derived recombinant mouse WIF-1 Gly29-Trp379 Accession # Q9WUA1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
11197
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9WUA1
WIF1, WIF-1, WNT inhibitory factor 1
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse WIF-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human WIF-1 is observed.
Mouse
Videos
Documents