missing translation for 'onlineSavingsMsg'
Learn More

WIF-1 Mouse anti-Human, Clone: 133015, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB134-500

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438565

  • £337.93 / 500µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

WIF-1 Monoclonal antibody MAB134500 Specifically detects WIF-1 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

WIF-1
133015
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WIF1
Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
11197
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
AAD25402
WIF1, WIF-1, WNT inhibitory factor 1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human WIF-1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse WIF-1 is observed.
Human
Videos