Learn More
WIF-1 Mouse anti-Human, Clone: 133009, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1342
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488555
  • £358.00 / 500µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
WIF-1 Monoclonal antibody MAB1342 Specifically detects WIF-1 in Human samples. It is validated for ELISA Capture
Specifications
Specifications
WIF-1
Unconjugated
Murine
IgG1
500μg
Primary
Human
133009
Purified
Mouse myeloma cell line NS0-derived recombinant human WIF-1. Gly29-Trp379
Protein A or G purified from hybridoma culture supernatant