Learn More
VSV-G Tag Polyclonal Antibody, HRP, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA126597
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-26597 detects VSV-G tag. PA1-26597 has been successfully used in and Western blot procedures. The PA1-26597 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues Y T D I E M N R L G K of VSV-G tag.
Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.Specifications
VSV-G Tag | |
Polyclonal | |
HRP | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Vesicular stomatitis virus; VSVG tag; VSV tag; VSV-G epitope tag; VSVG epitope tag; VSIVgp3; VSV epitope tag; YTDIEMNRLGK | |
Synthetic peptide: YTDIEMNRLGK conjugated to KLH | |
100 μg | |
Primary | |
Antibody | |
IgG |