missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSV-G Tag Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130138
258.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-30138 detects VSV-G from rabbit samples.
VSV-G Tag Polyclonal antibody specifically detects VSV-G Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
VSV-G Tag | |
Polyclonal | |
Unconjugated | |
vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; vsvg; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK | |
Rabbits were immunized with YTDIEMNRLGK (VSV-g epitope) conjugated with KLH. | |
100 μg | |
Primary | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4° C | |
Liquid | |
Tag |