Learn More
VRK1 Monoclonal Antibody (1F6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1828
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-828 detects VRK1 from human and mouse samples. MA1-828 has been successfully used in Western blot, immunofluorescence, and immunohistochemistry applications. For Western blotting, it is recommended to block with 3% BSA (not milk). The MA1-828 immunogen is recombinant fusion protein GST-VRK1.
VRK1 Monoclonal antibody specifically detects VRK1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
VRK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q80X41, Q99986 | |
VRK1 | |
Recombinant fusion protein GST-VRK1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1F6 | |
Unconjugated | |
VRK1 | |
51PK; MGC117401; MGC138280; PCH1; PCH1A; Serine/threonine-protein kinase 51PK; serine/threonine-protein kinase VRK1; vaccinia related kinase 1; vaccinia virus B1R-related kinase 1; vaccinia-related kinase 1; VRK1 | |
Mouse | |
Protein A | |
RUO | |
22367, 7443 | |
4° C | |
Liquid |