Learn More
VPS29 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530632
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HepG2. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (96%), Drosophila (82%), Xenopus laevis (97%), Dog (100%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VPS29 Polyclonal antibody specifically detects VPS29 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
VPS29 | |
Polyclonal | |
Unconjugated | |
VPS29 | |
2010015D08Rik; AW049835; DC15; DC7; DKFZp564F0223; EMBL:AAI67055.1, ECO:0000312; FLJ20492; hVPS29; MDS007; PEP11; PEP11 homolog; retromer protein; RGD:1308332}; UniProtKB:Q9UBQ0}; vacuolar protein sorting 29 (S. pombe); vacuolar protein sorting 29 homolog; vacuolar protein sorting 29 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 29; vacuolar protein sorting-associated protein 29 {ECO:0000250; vacuolar sorting protein 29; vacuolar sorting protein VPS29/PEP11; vesicle protein sorting 29; vesicle protein sorting 29 {ECO:0000250; VPS29; vps29 {ECO:0000312; VPS29 retromer complex component; VPS29, retromer complex component; x 007 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51699 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.78 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UBQ0 | |
VPS29 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 155 of Human VPS29 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |