missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VMAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA127477
Code : LD
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VMAT2 Polyclonal antibody specifically detects VMAT2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VMAT2 | |
Polyclonal | |
Unconjugated | |
SLC18A2 | |
1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214084, 25549, 6571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q01827, Q05940, Q8BRU6 | |
SLC18A2 | |
Purified VMAT protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |