Learn More
VLDL R Rat anti-Mouse, Clone: 268724, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2258
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429305
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
VLDLR Monoclonal antibody specifically detects VLDLR in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
VLDLR
268724
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
VLDLR
Mouse myeloma cell line NS0-derived recombinant mouse VLDL R Thr25-Ala798 Accession # P98156
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7436
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P98156
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL R, VLDL receptor, VLDL-R, VLDLRCH
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse VLDL R in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human ApoER2.