Learn More
Vitronectin Rat anti-Mouse, Clone: 347312, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3875
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460296
  • £271.93 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Vitronectin Monoclonal antibody MAB3875 Specifically detects Vitronectin in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Vitronectin
347312
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
VTN
Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478 Accession # AAH12690
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7448
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAH12690
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Vitronectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Vitronectin is observed.