missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GC Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518794
Code : LD
Additional Details : Weight : 0.01000kg
Description
GC Polyclonal antibody specifically detects GC in Human samples. It is validated for Western Blot.Specifications
GC | |
Polyclonal | |
Unconjugated | |
GC | |
DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2638 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P02774 | |
GC | |
Synthetic peptide sequence (CDNLSTKNSKFED) corresponding to the internal amino acids of GC | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |