Learn More
MCK-2 Goat anti-Viral, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF798
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440225
  • £289.09 / 100µg
Estimated Shipment
27-06-2019
Description
Description
MCK-2 Polyclonal antibody AF798 Specifically detects MCK-2 in Viral samples. It is validated for Western Blotting
Specifications
Specifications
MCK-2
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
E. coli-derived recombinant viral MCK-2
Immunogen affinity purified
RUO
Primary
Detects recombinant viral MCK-2 in direct ELISAs and Western blots.
Virus
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Reconstitute at 0.2 mg/mL in sterile PBS.