Learn More
Vinculin Monoclonal Antibody (SP117), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516424
Code : PN
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is heart or small intestine tissue.
Vinculin Monoclonal antibody specifically detects Vinculin in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Vinculin | |
Monoclonal | |
Unconjugated | |
VCL | |
9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
Rabbit | |
Protein A/G | |
RUO | |
7414 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
SP117 | |
PBS with 1% BSA and <0.1% sodium azide; pH 7.6 | |
P18206 | |
VCL | |
Synthetic peptide derived from C-terminus of human vinculin protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |