Learn More
Vimentin Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA514435
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14435 targets Vimentin in IHC (P) applications and shows reactivity with Human samples. The MA5-14435 immunogen is vimentin - recombinant protein encoding human Vimentin, gp100 - Extract of pigmented melanoma metastases from lymph nodes. Host/Isotype: Rabbit/Mouse / IgG/IgG1, kappa
Vimentin Cocktail antibody specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Vimentin | |
Cocktail | |
Unconjugated | |
Vim | |
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
Mixed | |
purified | |
RUO | |
7431 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Vimentin-SP20, gp100-HMB45, gp100-HMB45 | |
proprietary buffer with <0.1% sodium azide | |
P08670 | |
Vim | |
Vimentin - recombinant protein encoding human Vimentin, gp100 - Extract of pigmented melanoma metastases from lymph nodes | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG |