missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513300
Code : LE
Additional Details : Weight : 0.01000kg
Description
VEGFD Polyclonal antibody specifically detects VEGFD in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
VEGFD | |
Polyclonal | |
Unconjugated | |
VEGFD | |
AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2277 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
O43915 | |
VEGFD | |
KLH conjugated synthetic peptide between 322-352 amino acids from the C-terminal region of human VEGFD (VEGF4) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |