missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF R1/Flt-1 Rat anti-Mouse, Clone: 141515, R&D Systems™
Rat Monoclonal Antibody has been used in 6 publications
POA
Specifications
Antigen | VEGFR1/Flt-1 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1, Ser27-Glu759, Accession # P35969 |
Clone | 141515 |
Applications | Western Blot, Immunohistochemistry |
Conjugate | Unconjugated |
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VEGFR1/Flt-1 | |
141515 | |
Unconjugated | |
Rat | |
Primary | |
Detects mouse VEGFR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human VEGFR1, recombinant mouse (rm) VEGFR2, or rmVEGFR3 is observed. | |
Protein A or G purified from hybridoma culture supernatant |
Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1, Ser27-Glu759, Accession # P35969 | |
Western Blot, Immunohistochemistry | |
P35969 | |
IgG2a | |
2321 | |
RUO |
For Research Use Only