Learn More
VEGF-D Rat anti-Mouse, Clone: 90409, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB469
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450676
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
VEGF-D Monoclonal antibody MAB469 Specifically detects VEGF-D in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
VEGF-D
90409
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P97946
c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse VEGF-D in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) VEGF 165 rmVEGF-B 186 recombinant human (rh) VEGF-C, or rhVEGF-D is observed.
Mouse
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FIGF
E. coli-derived recombinant mouse VEGF-D Phe98-Ser206 Accession # P97946
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2277
Reconstitute at 0.5 mg/mL in sterile PBS.