missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF 165b Mouse anti-Human, Clone: 56-1, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
POA
Specifications
Antigen | VEGF |
---|---|
Clone | 56-1 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Conjugate | Unconjugated |
Form | Supernatant |
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
VEGF | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Supernatant | |
RUO | |
7422 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
56-1 | |
Unconjugated | |
Mouse | |
Human | |
KLH-conjugated human VEGF 165b synthetic peptide TCRSLTRKD Accession # AAL27435 | |
Primary |
For Research Use Only