missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDP Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518440
Code : A0
Additional Details : Weight : 0.01000kg
Description
VDP Polyclonal antibody specifically detects VDP in Human samples. It is validated for Western Blot.Specifications
VDP | |
Polyclonal | |
Unconjugated | |
USO1 | |
115kDa; General vesicular transport factor p115; P115; protein USO1 homolog; TAP; transcytosis associated protein; transcytosis associated protein p115; transcytosis-associated protein; USO1; USO1 homolog, vesicle docking protein; USO1 vesicle docking factor; USO1 vesicle docking protein homolog; USO1 vesicle transport factor; Vdp; VDP p115; vesicle docking protein p115; vesicle docking protein, 115 kDa; vesicle-docking protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8615 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O60763 | |
USO1 | |
Synthetic peptide sequence (DESEDPGKDLDH) corresponding to the C-terminus amino acids of USO1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |