Learn More
VDAC Recombinant Polyclonal Antibody (17HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710606
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
VDAC Recombinant Polyclonal antibody specifically detects VDAC in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
VDAC | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AL033343; BR1-VDAC; Brain-derived voltage-dependent anion channel 1; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; plasmalemmal porin; POR1; PORIN; Porin 31HL; porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; VDAC1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C | |
Recombinant protein corresponding to amino acids 150–250 of human Mitochondrial Porin | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Western Blot | |
17HCLC | |
Unconjugated | |
P21796 | |
Rabbit | |
Protein A | |
RUO | |
7416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |