Learn More
Vaccinia Virus Free form Monoclonal Antibody (I84B), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17484
Code : LD
Additional Details : Weight : 0.01000kg
Description
The MA1-7484 antibody reacts with Vaccinia virus, free form in viral and infected tissue samples. MA1-7484 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7484 antibody was raised against Vaccinia viral preparation.
Vaccinia Virus Free form Monoclonal antibody specifically detects Vaccinia Virus Free form in Virus samples. It is validated for ELISA, Immunocytochemistry.Specifications
Vaccinia Virus Free form | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Vaccinia viral preparation. | |
100 μg | |
Primary | |
4° C | |
Liquid |
ELISA, Immunocytochemistry | |
I84B | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |