missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VN1R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534310
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%).
VN1R1 Polyclonal antibody specifically detects VN1R1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
VN1R1 | |
Polyclonal | |
Unconjugated | |
VN1R1 | |
G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9GZP7 | |
VN1R1 | |
Synthetic 16 amino acid peptide from C-terminus of human VN1R1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |