Learn More
UNC5H2/UNC5B Mouse anti-Rat, Clone: 187305, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1006
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458245
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
UNC5H2/UNC5B Monoclonal antibody MAB1006 Specifically detects UNC5H2/UNC5B in Rat samples. It is validated for Western Blotting
Specifications
Specifications
UNC5H2/UNC5B
187305
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
UNC5B
Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
219699
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O08722
p53RDL1, p53-regulated receptor for death and life protein 1, Protein unc-5 homolog 2, Protein unc-5 homolog B, transmembrane receptor Unc5H2, unc5 (C.elegans homolog) b, unc-5 homolog 2, unc-5 homolog B (C. elegans), UNC5B, UNC5H2netrin receptor UNC5B
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat UNC5H2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat UNC5H1 or recombinant human UNC5H4 is observed.
Rat