Learn More
UNC13B Mouse anti-Human, Clone: 584102, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB6188
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461256
  • £249.49 / 100µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
UNC13B Monoclonal antibody MAB6188 Specifically detects UNC13B in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
UNC13B
584102
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
UNC13B
E. Coli-derived recombinant human UNC13B Asp418-Ser1591 Accession # O14795
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10497
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
O14795
hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, MUNC13B, protein unc-13 homolog B, unc-13 homolog B (C. elegans), Unc13h2, unc-13-like (C. elegans), UNC13Unc13h2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human UNC13B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) UNC13A and rhUNC13C is observed.