missing translation for 'onlineSavingsMsg'
Learn More

UGRP1/SCGB3A2 Rat anti-Mouse, Clone: 381707, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3465

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440086

  • £249.49 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

UGRP1/SCGB3A2 Monoclonal antibody MAB3465 Specifically detects UGRP1/SCGB3A2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

UGRP1/SCGB3A2
381707
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SCGB3A2
E. coli-derived recombinant mouse UGRP1/SCGB3A2 isoform C Leu22-Leu139 Accession # Q920H1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
117156
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q920H1
LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse UGRP1/SCGB3A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human UGRP1 is observed.
Mouse
Videos

For Research Use Only