missing translation for 'onlineSavingsMsg'
Learn More

TWEAK R/TNFRSF12 Rat anti-Mouse, Clone: 314636, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB16101

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448775

  • £288.60 / 100µg

Estimated Shipment
15-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TWEAK R/TNFRSF12 Monoclonal antibody MAB16101 Specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

TWEAK R/TNFRSF12
314636
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFRSF12A
E. coli-derived recombinant mouse TWEAK R Glu28-Trp79 Accession # Q9CR75.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
51330
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q9CR75.1
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TWEAK R/TNFRSF12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) EDA, rmNeogenin, recombinant human TWEAK R, rmOPG, or rmTNF R1 is observed.
Mouse
Videos
Documents

For Research Use Only