Learn More
TWEAK R/TNFRSF12 Rat anti-Human, Clone: 314502, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1199
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488435
  • £275.00 / 100µg
Estimated Shipment
08-03-2018
Add to basket
Description
Description
TWEAK R/TNFRSF12 Monoclonal antibody MAB1199 Specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blotting
Specifications
Specifications
TWEAK R/TNFRSF12
Unconjugated
Rat
IgG2a
100μg
Primary
Human
314502
Purified
E.Coli-derived recombinant human TWEAK R/TNFRSF12. Glu28-Trp79
Protein A or G purified from hybridoma culture supernatant