Learn More
TSP50 Rat anti-Mouse, Clone: 260514, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2455
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469455
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
TSP50 Monoclonal antibody MAB2455 Specifically detects TSP50 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
TSP50
260514
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_666339.2
Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TSP50 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human TSP50 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Prss50
Mouse myeloma cell line NS0-derived recombinant mouse TSP50 His58-Leu439 Accession # NP_666339.2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
29122
Reconstitute at 0.5 mg/mL in sterile PBS.