missing translation for 'onlineSavingsMsg'
Learn More

TSKS, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA116660

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11545353

  • £257.00 / 100┬ÁL


Estimated Shipment
20-06-2018
Add to basket

Specifications

Specifications

TSKS
Polyclonal
whole serum with 0.02% sodium azide
Q9UJT2
STK22S1, TSKS1, TSSKS
Rabbit
IgG
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
60385
Western Blot
Unconjugated
TSKS
Serum
TSKS
Residues 577-592 of the human TSKS protein
100μL
Antibody
Polyclonal
Human
Videos