Learn More
TSG Rat anti-Mouse, Clone: 120024, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB756
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441816
  • £337.93 / 500µg
Estimated Shipment date
from Supplier 25-09-2019
Description
Description
TSG Monoclonal antibody MAB756 Specifically detects TSG in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
TSG
120024
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Q9EP52
TSGtwisted gastrulation protein homolog 1, twisted gastrulation homolog 1 (Drosophila), TWSG1
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TSG in direct ELISAs and Western blots.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TWSG1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSG Cys25-Phe222 Accession # Q9EP52
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
57045
Reconstitute at 0.5 mg/mL in sterile PBS.