Learn More
Tryptase gamma-1/TPSG1 Rat anti-Human, Clone: 279005, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1667
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438815
  • £249.49 / 100µg
Estimated Shipment
28-06-2019
Description
Description
Tryptase gamma-1/TPSG1 Monoclonal antibody MAB1667 Specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Tryptase gamma-1/TPSG1
279005
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TPSG1
Mouse myeloma cell line NS0-derived recombinant human Tryptase gamma-1/TPSG1 Arg20-Arg281 Accession # Q9NRR2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
25823
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q9NRR2
EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, TPSG1, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II, Tryptase gamma1
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Tryptase gamma-1/TPSG1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCPT-1, recombinant human (rh) Tryptase alpha , rmTryptase beta-1, rhTryptase beta-2, or rmTryptase-5 is observed.