Learn More
Tryptase epsilon/BSSP-4 Rat anti-Mouse, Clone: 246702, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB20592
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449135
  • £376.32 / 500µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Tryptase epsilon/BSSP-4 Monoclonal antibody MAB20592 Specifically detects Tryptase epsilon/BSSP-4 in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
Tryptase epsilon/BSSP-4/PRSS22
246702
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9ER10
BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Tryptase epsilon/BSSP-4 in ELISAs.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PRSS22
Mouse myeloma cell line NS0-derived recombinant mouse Tryptase epsilon/BSSP-4 Ala33-Ser306 Accession # Q9ER10
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
64063
Reconstitute at 0.5 mg/mL in sterile PBS.