Learn More
Tryptase beta-1/MCP-7/Mcpt7 Rat anti-Mouse, Clone: 263019, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1937
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469045
  • £427.77 / 500µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Tryptase beta-1/MCP-7/Mcpt7 Monoclonal antibody MAB1937 Specifically detects Tryptase beta-1/MCP-7/Mcpt7 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Tryptase beta-1/MCP-7/Mcpt7
263019
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q02844
Tpsab1
Mouse myeloma cell line NS0-derived recombinant mouse Tryptase beta-1/MCP-7/Mcpt7 Ile29-Phe273 Accession # Q02844
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects mouse Tryptase beta-1/MCP-7/Mcpt7 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Tryptase alpha and rhTryptase beta-2 is observed and no cross-reactivity with recombinant mouse MCPT-1 is observed.