Learn More
Trypsin 1/PRSS1 Mouse anti-Human, Clone: 446628, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3848
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440286
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 19-12-2019
Description
Description
Trypsin 1/PRSS1 Monoclonal antibody MAB3848 Specifically detects Trypsin 1/PRSS1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Trypsin 1/PRSS1
446628
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PRSS1
Mouse myeloma cell line NS0-derived recombinant human Trypsin 1/PRSS1 Ala16-Ser247 Accession # P07477
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5644
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P07477
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), PRSS1, PTRYI, Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, Trygn16, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Trpysin 1/PRSS1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) Trypsin 3, recombinant human (rh) Trypsin 2, or rhTrypsin 3.