missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OST00129W
Code : LD
Additional Details : Weight : 0.99000kg
Description
Glycerol (1:1) may be added for additional stability.
Specifications
TRP | |
Polyclonal | |
trp | |
Transient receptor potential cation channel protein | |
Rabbit | |
100 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P19334 | |
TRP | |
A synthetic peptide from the 4th cytoplasmic loop of drosophila TRP | |
RUO | |
43542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |