Learn More
TPM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522012
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (84%), Pig (98%), Rabbit (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPM2 Polyclonal antibody specifically detects TPM2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TPM2 | |
Polyclonal | |
Unconjugated | |
TPM2 | |
AMCD1; beta tropomyosin; beta-tropomyosin; cb836; cytoskeletal protein; DA1; DA2B; epididymis secretory protein Li 273; fj63f03; HEL-S-273; NEM4; nemaline myopathy type 4; RP11-112J3.4; TMSB; TPM2; Tpm-2; tpm4l; Trop-2; tropomyosin; tropomyosin 2; tropomyosin 2 (beta); tropomyosin 2, beta; tropomyosin beta chain; Tropomyosin-2; unnamed protein product; wu:fj63f03; zgc:86810 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.66 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P07951 | |
TPM2 | |
Full length human Tropomyosin 2 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |