missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531491
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRIM4 Polyclonal antibody specifically detects TRIM4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TRIM4 | |
Polyclonal | |
Unconjugated | |
TRIM4 | |
E3 ubiquitin-protein ligase TRIM4; RING finger protein 87; RING-type E3 ubiquitin transferase TRIM4; RNF87; TRIM4; tripartite motif containing 4; tripartite motif protein 4; tripartite motif-containing 4; Tripartite motif-containing protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9C037 | |
TRIM4 | |
Recombinant protein fragment corresponding to a region within amino acids 79 and 268 of Human TRIM4/ RNF87 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |