missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518155
Code : A0
Additional Details : Weight : 0.01000kg
Description
TRIM4 Polyclonal antibody specifically detects TRIM4 in Human samples. It is validated for Western Blot.Specifications
TRIM4 | |
Polyclonal | |
Unconjugated | |
TRIM4 | |
E3 ubiquitin-protein ligase TRIM4; RING finger protein 87; RING-type E3 ubiquitin transferase TRIM4; RNF87; TRIM4; tripartite motif containing 4; tripartite motif protein 4; tripartite motif-containing 4; Tripartite motif-containing protein 4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
89122 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9C037 | |
TRIM4 | |
Synthetic peptide sequence (ASLVIPPVTDRK) corresponding to the C-terminus amino acids of TRIM4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |